Learn More
STEMCELL Technologies MEM with Earle's & NEAA, Size: 500 mL
SDP
Catalog No. 501974758
Encompass
Minimum Essential Medium Eagle with Earle's salts and non-essential amino acids (NEAA) is recommended for a wide variety of cell culture applications. Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition.
Catalog No. 50-197-4758 Supplier STEMCELL Technologies Supplier No. 36550
Minimum Essential Medium Eagle with Earle's salts and non-essential amino acids (NEAA) is recommended for a wide variety of cell culture applications. Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition.