Learn More
Medchemexpress LLC Memantine-d6 (hydrochloride) | 1189713-18-5 | 99.8% | 500 UG
SDP
Catalog No. 5000406116
Encompass_Preferred
Memantine-d6 (hydrochloride) is a deuterium labeled Memantine hydrochloride. It functions as a moderate affinity, uncompetitive NMDA receptor antagonist, inhibiting CYP2B6 and CYP2D6. This compound is intended for research use only, primarily as a tracer and internal standard for quantitative analysis. Deuteration, involving the incorporation of stable heavy isotopes into drug molecules, is utilized for quantitation during drug development and may influence pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06116 Supplier Medchemexpress LLC Supplier No. HYB0365AS500UG
Add to Cart
Add to Cart
Memantine-d6 (hydrochloride) is a deuterium labeled Memantine hydrochloride. It functions as a moderate affinity, uncompetitive NMDA receptor antagonist, inhibiting CYP2B6 and CYP2D6. This compound is intended for research use only, primarily as a tracer and internal standard for quantitative analysis. Deuteration, involving the incorporation of stable heavy isotopes into drug molecules, is utilized for quantitation during drug development and may influence pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules.
  • Deuteration has the potential to affect pharmacokinetic and metabolic profiles of drugs.