Learn More
Enzo Life Sciences Memantine . hydrochloride (50mg). CAS: 41100-52-1
SDP
Supplier:  Enzo Life Sciences ALX550363M050
Encompass_Preferred
Low-affinity NMDA receptor antagonist. Stimulates dopamine release. Alternative name: 1-Amino-3,5-dimethyladamantane . HCl, 3,5-Dimethyl-1-aminoadamantane . HCl. Purity: ≥98% (Argentmetric Titration). Appearance: White powder. Long Term Storage: +4°C.
Catalog No. 50-200-9546