Learn More
Apexbio Technology LLC Memantine hydrochloride, 5g Cas# 41100-52-1
SDP
Supplier:  Apexbio Technology LLC B33085000
Encompass_Preferred
NMDA receptor antagonist. Memantine hydrochloride. 1401033-86-0. MFCD28155269 Memantine hydrochloride is an antagonist of NMDA receptor with IC50 values of 0.93M, 0.82M and 0.47M for NRIa/NR2A, NRIa/NR2B and NRIa/NR2D, respectively [1]. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8701