Learn More
Hampton Research MEMBFAC NO01 185ML
SDP
Catalog No. 501752
Encompass
HR2-920-01/MembFac #01 (A1), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.6, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-175-2 Supplier Hampton Research Supplier No. HR292001
HR2-920-01/MembFac #01 (A1), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.6, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.