Learn More
Hampton Research MEMBFAC NO02 185ML
SDP
Catalog No. 501753
Encompass
HR2-920-02/MembFac #02 (A2), 185 ml - 0.1 M Zinc acetate dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 12% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-175-3 Supplier Hampton Research Supplier No. HR292002
HR2-920-02/MembFac #02 (A2), 185 ml - 0.1 M Zinc acetate dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 12% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.