Learn More
Hampton Research MEMBFAC NO03 185ML
SDP
Catalog No. 501754
Encompass
HR2-920-03/MembFac #03 (A3), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-175-4 Supplier Hampton Research Supplier No. HR292003
HR2-920-03/MembFac #03 (A3), 185 ml - 0.2 M Ammonium sulfate, 0.1 M Sodium acetate trihydrate pH 4.6, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.