Learn More
Hampton Research MEMBFAC NO04 185ML
SDP
Catalog No. 501755
Encompass
HR2-920-04/MembFac #04 (A4), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.6, 12% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-175-5 Supplier Hampton Research Supplier No. HR292004
HR2-920-04/MembFac #04 (A4), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.6, 12% v/v 2-Propanol. Please note items from this supplier are non-returnable.