Learn More
Hampton Research MEMBFAC NO05 185ML
SDP
Catalog No. 501756
Encompass
HR2-920-05/MembFac #05 (A5), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 12% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-175-6 Supplier Hampton Research Supplier No. HR292005
HR2-920-05/MembFac #05 (A5), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 12% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.