Learn More
Hampton Research MEMBFAC NO06 185ML
SDP
Catalog No. 501757
Encompass
HR2-920-06/MembFac #06 (A6), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-175-7 Supplier Hampton Research Supplier No. HR292006
HR2-920-06/MembFac #06 (A6), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.