Learn More
Hampton Research MEMBFAC NO07 185ML
SDP
Catalog No. 501758
Encompass
HR2-920-07/MembFac #07 (A7), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Magnesium sulfate heptahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-175-8 Supplier Hampton Research Supplier No. HR292007
HR2-920-07/MembFac #07 (A7), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Magnesium sulfate heptahydrate. Please note items from this supplier are non-returnable.