Learn More
Hampton Research MEMBFAC NO08 185ML
SDP
Catalog No. 501759
Encompass
HR2-920-08/MembFac #08 (A8), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-175-9 Supplier Hampton Research Supplier No. HR292008
HR2-920-08/MembFac #08 (A8), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.