Learn More
Hampton Research MEMBFAC NO09 185ML
SDP
Catalog No. 501760
Encompass
HR2-920-09/MembFac #09 (A9), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.
Catalog No. 50-176-0 Supplier Hampton Research Supplier No. HR292009
HR2-920-09/MembFac #09 (A9), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Ammonium phosphate monobasic. Please note items from this supplier are non-returnable.