Learn More
Hampton Research MEMBFAC NO10 185ML
SDP
Catalog No. 501761
Encompass
HR2-920-10/MembFac #10 (A10), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.6, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-176-1 Supplier Hampton Research Supplier No. HR292010
HR2-920-10/MembFac #10 (A10), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium acetate trihydrate pH 4.6, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.