Learn More
Hampton Research MEMBFAC NO11 185ML
SDP
Catalog No. 501762
Encompass
HR2-920-11/MembFac #11 (A11), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-176-2 Supplier Hampton Research Supplier No. HR292011
HR2-920-11/MembFac #11 (A11), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.