Learn More
Hampton Research MEMBFAC NO12 185ML
SDP
Catalog No. 501763
Encompass
HR2-920-12/MembFac #12 (A12), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-176-3 Supplier Hampton Research Supplier No. HR292012
HR2-920-12/MembFac #12 (A12), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.