Learn More
Hampton Research MEMBFAC NO13 185ML
SDP
Catalog No. 501764
Encompass
HR2-920-13/MembFac #13 (B1), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 12% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-176-4 Supplier Hampton Research Supplier No. HR292013
HR2-920-13/MembFac #13 (B1), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 12% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.