Learn More
Hampton Research MEMBFAC NO14 185ML
SDP
Catalog No. 501765
Encompass
HR2-920-14/MembFac #14 (B2), 185 ml - 0.1 M Sodium citrate tribasic dihydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 10% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-176-5 Supplier Hampton Research Supplier No. HR292014
HR2-920-14/MembFac #14 (B2), 185 ml - 0.1 M Sodium citrate tribasic dihydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 10% v/v 2-Propanol. Please note items from this supplier are non-returnable.