Learn More
Hampton Research MEMBFAC NO15 185ML
SDP
Catalog No. 501766
Encompass
HR2-920-15/MembFac #15 (B3), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-176-6 Supplier Hampton Research Supplier No. HR292015
HR2-920-15/MembFac #15 (B3), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.