Learn More
Hampton Research MEMBFAC NO16 185ML
SDP
Catalog No. 501767
Encompass
HR2-920-16/MembFac #16 (B4), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 1.0 M Magnesium sulfate heptahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-176-7 Supplier Hampton Research Supplier No. HR292016
HR2-920-16/MembFac #16 (B4), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 1.0 M Magnesium sulfate heptahydrate. Please note items from this supplier are non-returnable.