Learn More
Hampton Research MEMBFAC NO17 185ML
SDP
Catalog No. 501768
Encompass
HR2-920-17/MembFac #17 (B5), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 12% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-176-8 Supplier Hampton Research Supplier No. HR292017
HR2-920-17/MembFac #17 (B5), 185 ml - 0.1 M Sodium chloride, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 12% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.