Learn More
Hampton Research MEMBFAC NO18 185ML
SDP
Catalog No. 501769
Encompass
HR2-920-18/MembFac #18 (B6), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-176-9 Supplier Hampton Research Supplier No. HR292018
HR2-920-18/MembFac #18 (B6), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.