Learn More
Hampton Research MEMBFAC NO19 185ML
SDP
Catalog No. 501770
Encompass
HR2-920-19/MembFac #19 (B7), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 4% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-177-0 Supplier Hampton Research Supplier No. HR292019
HR2-920-19/MembFac #19 (B7), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 4% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.