Learn More
Hampton Research MEMBFAC NO20 185ML
SDP
Catalog No. 501771
Encompass
HR2-920-20/MembFac #20 (B8), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 0.1 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-177-1 Supplier Hampton Research Supplier No. HR292020
HR2-920-20/MembFac #20 (B8), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 0.1 M Sodium chloride. Please note items from this supplier are non-returnable.