Learn More
Hampton Research MEMBFAC NO21 185ML
SDP
Catalog No. 501772
Encompass
HR2-920-21/MembFac #21 (B9), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 4% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-177-2 Supplier Hampton Research Supplier No. HR292021
HR2-920-21/MembFac #21 (B9), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 4% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.