Learn More
Hampton Research MEMBFAC NO23 185ML
SDP
Catalog No. 501774
Encompass
HR2-920-23/MembFac #23 (B11), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M ADA pH 6.5, 12% w/v Polyethylene glycol 4,000, 2% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-177-4 Supplier Hampton Research Supplier No. HR292023
HR2-920-23/MembFac #23 (B11), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M ADA pH 6.5, 12% w/v Polyethylene glycol 4,000, 2% v/v 2-Propanol. Please note items from this supplier are non-returnable.