Learn More
Hampton Research MEMBFAC NO24 185ML
SDP
Catalog No. 501775
Encompass
HR2-920-24/MembFac #24 (B12), 185 ml - 0.1 M ADA pH 6.5, 1.0 M Ammonium phosphate dibasic. Please note items from this supplier are non-returnable.
Catalog No. 50-177-5 Supplier Hampton Research Supplier No. HR292024
HR2-920-24/MembFac #24 (B12), 185 ml - 0.1 M ADA pH 6.5, 1.0 M Ammonium phosphate dibasic. Please note items from this supplier are non-returnable.