Learn More
Hampton Research MEMBFAC NO25 185ML
SDP
Catalog No. 501776
Encompass
HR2-920-25/MembFac #25 (C1), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M ADA pH 6.5, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-177-6 Supplier Hampton Research Supplier No. HR292025
HR2-920-25/MembFac #25 (C1), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M ADA pH 6.5, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.