Learn More
Hampton Research MEMBFAC NO26 185ML
SDP
Catalog No. 501777
Encompass
HR2-920-26/MembFac #26 (C2), 185 ml - 0.1 M ADA pH 6.5, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-177-7 Supplier Hampton Research Supplier No. HR292026
HR2-920-26/MembFac #26 (C2), 185 ml - 0.1 M ADA pH 6.5, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.