Learn More
Hampton Research MEMBFAC NO27 185ML
SDP
Catalog No. 501778
Encompass
HR2-920-27/MembFac #27 (C3), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M ADA pH 6.5, 1.0 M Magnesium sulfate hydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-177-8 Supplier Hampton Research Supplier No. HR292027
HR2-920-27/MembFac #27 (C3), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M ADA pH 6.5, 1.0 M Magnesium sulfate hydrate. Please note items from this supplier are non-returnable.