Learn More
Hampton Research MEMBFAC NO28 185ML
SDP
Catalog No. 501779
Encompass
HR2-920-28/MembFac #28 (C4), 185 ml - 0.3 M Lithium sulfate monohydrate, 0.1 M ADA pH 6.5, 4% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-177-9 Supplier Hampton Research Supplier No. HR292028
HR2-920-28/MembFac #28 (C4), 185 ml - 0.3 M Lithium sulfate monohydrate, 0.1 M ADA pH 6.5, 4% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.