Learn More
Hampton Research MEMBFAC NO29 185ML
SDP
Catalog No. 501780
Encompass
HR2-920-29/MembFac #29 (C5), 185 ml - 0.1 M Ammonium sulfate, 0.1 M HEPES sodium pH 7.5, 0.5 M Sodium phosphate dibasic dihydrate, 0.5 M Potassium phosphate dibasic. Please note items from this supplier are non-returnable.
Catalog No. 50-178-0 Supplier Hampton Research Supplier No. HR292029
HR2-920-29/MembFac #29 (C5), 185 ml - 0.1 M Ammonium sulfate, 0.1 M HEPES sodium pH 7.5, 0.5 M Sodium phosphate dibasic dihydrate, 0.5 M Potassium phosphate dibasic. Please note items from this supplier are non-returnable.