Learn More
Hampton Research MEMBFAC NO30 185ML
SDP
Catalog No. 501781
Encompass
HR2-920-30/MembFac #30 (C6), 185 ml - 0.1 M Sodium chloride, 0.1 M HEPES sodium pH 7.5, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-178-1 Supplier Hampton Research Supplier No. HR292030
HR2-920-30/MembFac #30 (C6), 185 ml - 0.1 M Sodium chloride, 0.1 M HEPES sodium pH 7.5, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.