Learn More
Hampton Research MEMBFAC NO31 185ML
SDP
Catalog No. 501782
Encompass
HR2-920-31/MembFac #31 (C7), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-178-2 Supplier Hampton Research Supplier No. HR292031
HR2-920-31/MembFac #31 (C7), 185 ml - 0.1 M Magnesium chloride hexahydrate, 0.1 M HEPES sodium pH 7.5, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.