Learn More
Hampton Research MEMBFAC NO32 185ML
SDP
Catalog No. 501783
Encompass
HR2-920-32/MembFac #32 (C8), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.0 M Potassium sodium tartrate tetrahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-178-3 Supplier Hampton Research Supplier No. HR292032
HR2-920-32/MembFac #32 (C8), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.0 M Potassium sodium tartrate tetrahydrate. Please note items from this supplier are non-returnable.