Learn More
Hampton Research MEMBFAC NO33 185ML
SDP
Catalog No. 501784
Encompass
HR2-920-33/MembFac #33 (C9), 185 ml - 0.1 M Ammonium sulfate, 0.1 M HEPES sodium pH 7.5, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-178-4 Supplier Hampton Research Supplier No. HR292033
HR2-920-33/MembFac #33 (C9), 185 ml - 0.1 M Ammonium sulfate, 0.1 M HEPES sodium pH 7.5, 18% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.