Learn More
Hampton Research MEMBFAC NO34 185ML
SDP
Catalog No. 501785
Encompass
HR2-920-34/MembFac #34 (C10), 185 ml - 0.1 M Ammonium sulfate, 0.1 M HEPES sodium pH 7.5, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-178-5 Supplier Hampton Research Supplier No. HR292034
HR2-920-34/MembFac #34 (C10), 185 ml - 0.1 M Ammonium sulfate, 0.1 M HEPES sodium pH 7.5, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.