Learn More
Hampton Research MEMBFAC NO35 185ML
SDP
Catalog No. 501786
Encompass
HR2-920-35/MembFac #35 (C11), 185 ml - 0.1 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-178-6 Supplier Hampton Research Supplier No. HR292035
HR2-920-35/MembFac #35 (C11), 185 ml - 0.1 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.