Learn More
Hampton Research MEMBFAC NO36 185ML
SDP
Catalog No. 501787
Encompass
HR2-920-36/MembFac #36 (C12), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.0 M Sodium citrate tribasic dihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-178-7 Supplier Hampton Research Supplier No. HR292036
HR2-920-36/MembFac #36 (C12), 185 ml - 0.1 M HEPES sodium pH 7.5, 1.0 M Sodium citrate tribasic dihydrate. Please note items from this supplier are non-returnable.