Learn More
Hampton Research MEMBFAC NO37 185ML
SDP
Catalog No. 501788
Encompass
HR2-920-37/MembFac #37 (D1), 185 ml - 0.6 M Magnesium sulfate hydrate, 0.1 M HEPES sodium pH 7.5, 4% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-178-8 Supplier Hampton Research Supplier No. HR292037
HR2-920-37/MembFac #37 (D1), 185 ml - 0.6 M Magnesium sulfate hydrate, 0.1 M HEPES sodium pH 7.5, 4% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.