Learn More
Hampton Research MEMBFAC NO38 185ML
SDP
Catalog No. 501789
Encompass
HR2-920-38/MembFac #38 (D2), 185 ml - 0.6 M Magnesium sulfate hydrate, 0.1 M HEPES sodium pH 7.5, 4% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-178-9 Supplier Hampton Research Supplier No. HR292038
HR2-920-38/MembFac #38 (D2), 185 ml - 0.6 M Magnesium sulfate hydrate, 0.1 M HEPES sodium pH 7.5, 4% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.