Learn More
Hampton Research MEMBFAC NO39 185ML
SDP
Catalog No. 501790
Encompass
HR2-920-39/MembFac #39 (D3), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M HEPES sodium pH 7.5, 0.1 M Potassium sodium tartrate tetrahydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-179-0 Supplier Hampton Research Supplier No. HR292039
HR2-920-39/MembFac #39 (D3), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M HEPES sodium pH 7.5, 0.1 M Potassium sodium tartrate tetrahydrate. Please note items from this supplier are non-returnable.