Learn More
Hampton Research MEMBFAC NO40 185ML
SDP
Catalog No. 501791
Encompass
HR2-920-40/MembFac #40 (D4), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M TRIS hydrochloride pH 8.5, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-179-1 Supplier Hampton Research Supplier No. HR292040
HR2-920-40/MembFac #40 (D4), 185 ml - 0.1 M Lithium sulfate monohydrate, 0.1 M TRIS hydrochloride pH 8.5, 12% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.