Learn More
Hampton Research MEMBFAC NO41 185ML
SDP
Catalog No. 501792
Encompass
HR2-920-41/MembFac #41 (D5), 185 ml - 0.1 M Ammonium phosphate dibasic, 0.1 M TRIS hydrochloride pH 8.5, 0.5 M Sodium phosphate dibasic dihydrate, 0.5 M Potassium phosphate dibasic. Please note items from this supplier are non-returnable.
Catalog No. 50-179-2 Supplier Hampton Research Supplier No. HR292041
HR2-920-41/MembFac #41 (D5), 185 ml - 0.1 M Ammonium phosphate dibasic, 0.1 M TRIS hydrochloride pH 8.5, 0.5 M Sodium phosphate dibasic dihydrate, 0.5 M Potassium phosphate dibasic. Please note items from this supplier are non-returnable.