Learn More
Hampton Research MEMBFAC NO42 185ML
SDP
Catalog No. 501793
Encompass
HR2-920-42/MembFac #42 (D6), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 0.1 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-179-3 Supplier Hampton Research Supplier No. HR292042
HR2-920-42/MembFac #42 (D6), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 0.1 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.