Learn More
Hampton Research MEMBFAC NO43 185ML
SDP
Catalog No. 501794
Encompass
HR2-920-43/MembFac #43 (D7), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 0.1 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-179-4 Supplier Hampton Research Supplier No. HR292043
HR2-920-43/MembFac #43 (D7), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 0.1 M Sodium chloride. Please note items from this supplier are non-returnable.