Learn More
Hampton Research MEMBFAC NO44 185ML
SDP
Catalog No. 501795
Encompass
HR2-920-44/MembFac #44 (D8), 185 ml - 0.1 M Ammonium phosphate dibasic, 0.1 M TRIS hydrochloride pH 8.5, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.
Catalog No. 50-179-5 Supplier Hampton Research Supplier No. HR292044
HR2-920-44/MembFac #44 (D8), 185 ml - 0.1 M Ammonium phosphate dibasic, 0.1 M TRIS hydrochloride pH 8.5, 12% w/v Polyethylene glycol 6,000. Please note items from this supplier are non-returnable.