Learn More
Hampton Research MEMBFAC NO45 185ML
SDP
Catalog No. 501796
Encompass
HR2-920-45/MembFac #45 (D9), 185 ml - 0.1 M Potassium sodium tartrate tetrahydrate, 0.1 M TRIS hydrochloride pH 8.5, 0.4 M Magnesium sulfate hydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-179-6 Supplier Hampton Research Supplier No. HR292045
HR2-920-45/MembFac #45 (D9), 185 ml - 0.1 M Potassium sodium tartrate tetrahydrate, 0.1 M TRIS hydrochloride pH 8.5, 0.4 M Magnesium sulfate hydrate. Please note items from this supplier are non-returnable.