Learn More
Hampton Research MEMBFAC NO46 185ML
SDP
Catalog No. 501797
Encompass
HR2-920-46/MembFac #46 (D10), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 0.2 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-179-7 Supplier Hampton Research Supplier No. HR292046
HR2-920-46/MembFac #46 (D10), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 0.2 M Lithium sulfate monohydrate. Please note items from this supplier are non-returnable.