Learn More
Hampton Research MEMBFAC NO47 185ML
SDP
Catalog No. 501798
Encompass
HR2-920-47/MembFac #47 (D11), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 0.5 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-179-8 Supplier Hampton Research Supplier No. HR292047
HR2-920-47/MembFac #47 (D11), 185 ml - 0.1 M TRIS hydrochloride pH 8.5, 0.5 M Ammonium sulfate. Please note items from this supplier are non-returnable.